Assay Details
Target Gene Details
Entrez Gene ID: | 140707 |
Gene Name: | BRI3 binding protein |
Gene Aliases: |
BNAS1, HCCR-1, HCCR-2, HCCRBP-1, KG19 |
Location: |
Chr.12:124993648-125051845 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRI3BP | XM_011537940.2 | XP_011536242.1 | ||
| AF284094.1 | 3 | 1590 | AAL54382.1 | |
| AK055703.1 | 1 | 1167 |
Target Gene Details
Entrez Gene ID: | 102659353 |
Gene Name: | TNF and HNRNPL related immunoregulatory long non-coding RNA |
Gene Aliases: |
BRI3BP-AS1, BRI3BPAS1, Linc1992, TCONS_00020260 |
Location: |
Chr.12:125025443-125027423 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRIL | NR_110375.1 | 1 | 1247 | |
| AK025766.1 | 1 | 246 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047470 | Chr.12:125009120 - 125044073 on Build GRCh38 | Loss |
|
| nsv1051020 | Chr.12:125015140 - 125809096 on Build GRCh38 | Gain |
|
| esv3631032 | Chr.12:125022783 - 125033641 on Build GRCh38 | Gain |
|
| esv3892269 | Chr.12:125024464 - 125055560 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73231614] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map