Assay Details
Target Gene Details
Entrez Gene ID: | 56938 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like 2 |
Gene Aliases: |
BMAL2, CLIF, MOP9, PASD9, bHLHe6 |
Location: |
Chr.12:27332854-27425813 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Gene Details
Entrez Gene ID: | 101928646 |
Gene Name: | ARNTL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:27389789-27446634 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2-AS1 | NR_109975.1 | |||
| BU568793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
|
| dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
|
| esv3628933 | Chr.12:27275994 - 27403117 on Build GRCh38 | Gain |
|
| nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
|
| nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75144234] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map