Assay Details
Target Gene Details
Entrez Gene ID: | 64375 |
Gene Name: | IKAROS family zinc finger 4 |
Gene Aliases: |
EOS, ZNFN1A4 |
Location: |
Chr.12:56007647-56038435 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF4 | XM_005269086.3 | XP_005269143.1 | ||
| XM_005269090.3 | XP_005269147.1 | |||
| XM_011538664.1 | XP_011536966.1 | |||
| XM_011538666.2 | XP_011536968.1 | |||
| XM_011538669.2 | XP_011536971.1 | |||
| XM_017019805.1 | XP_016875294.1 | |||
| XM_017019806.1 | XP_016875295.1 | |||
| XM_017019807.1 | XP_016875296.1 | |||
| XM_017019809.1 | XP_016875298.1 | |||
| XM_017019810.1 | XP_016875299.1 | |||
| XM_017019812.1 | XP_016875301.1 | |||
| XM_017019813.1 | XP_016875302.1 | |||
| XM_017019814.1 | XP_016875303.1 | |||
| XM_017019815.1 | XP_016875304.1 | |||
| XM_017019816.1 | XP_016875305.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
| nsv832427 | Chr.12:55998761 - 56118990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map