Assay Details
Target Gene Details
Entrez Gene ID: | 29953 |
Gene Name: | thyrotropin releasing hormone degrading enzyme |
Gene Aliases: |
PAP-II, PGPEP2, TRH-DE |
Location: |
Chr.12:72087288-72666662 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRHDE | XM_017019244.1 | XP_016874733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518498 | Chr.12:71913836 - 72114999 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map