Assay Details
Target Gene Details
Entrez Gene ID: | 10728 |
Gene Name: | prostaglandin E synthase 3 |
Gene Aliases: |
P23, TEBP, cPGES |
Location: |
Chr.12:56663341-56689250 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES3 | XM_005268576.4 | 1 | 591 | XP_005268633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map