Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832511 | Chr.12:106551876 - 106714050 on Build GRCh38 | Gain |
|
| nsv832510 | Chr.12:106393952 - 106557362 on Build GRCh38 | Loss |
|
| nsv560090 | Chr.12:106541358 - 106830191 on Build GRCh38 | Gain |
|

Genomic Map