Assay Details
Target Gene Details
Entrez Gene ID: | 56994 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
CPT, CPT1 |
Location: |
Chr.12:101697639-101752038 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHPT1 |
Target Gene Details
Entrez Gene ID: | 79158 |
Gene Name: | N-acetylglucosamine-1-phosphate transferase alpha and beta subunits |
Gene Aliases: |
GNPTA, ICD |
Location: |
Chr.12:101745497-101830867 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNPTAB | NM_024312.4 | NP_077288.2 | ||
| XM_011538731.2 | XP_011537033.1 | |||
| XM_017019961.1 | XP_016875450.1 | |||
| XM_017019962.1 | XP_016875451.1 | |||
| AB033034.2 | BAA86522.2 | |||
| AK001821.1 | BAA91926.1 | |||
| AL359588.1 | CAB94874.1 | |||
| AM085438.1 | CAJ30014.1 | |||
| AY687932.1 | AAV98624.1 | |||
| BC042615.1 | AAH42615.1 | |||
| BC131787.1 | ||||
| CR749871.1 | CAH18711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832497 | Chr.12:101667198 - 101861862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map