Assay Details
Target Gene Details
Entrez Gene ID: | 84102 |
Gene Name: | solute carrier family 41 member 2 |
Gene Aliases: |
SLC41A1-L1 |
Location: |
Chr.12:104802553-104958744 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC41A2 | NM_032148.3 | NP_115524.3 | ||
| XM_005269176.1 | XP_005269233.1 | |||
| XM_005269178.1 | XP_005269235.1 | |||
| XM_005269179.1 | XP_005269236.1 | |||
| XM_006719630.2 | XP_006719693.1 | |||
| XM_011538807.2 | XP_011537109.1 | |||
| XM_011538808.1 | XP_011537110.1 | |||
| XM_011538809.1 | XP_011537111.1 | |||
| XM_011538810.2 | XP_011537112.1 | |||
| XM_011538811.2 | XP_011537113.1 | |||
| XM_011538813.2 | XP_011537115.1 | |||
| XM_017020013.1 | XP_016875502.1 | |||
| XM_017020014.1 | XP_016875503.1 | |||
| XM_017020016.1 | XP_016875505.1 | |||
| XM_017020017.1 | XP_016875506.1 | |||
| AK027838.1 | BAB55402.1 | |||
| AL136828.1 | CAB66762.1 | |||
| BC036734.1 | AAH36734.1 | |||
| BC106871.1 | ||||
| BC106872.1 |
More Information
Set Membership: |
|

Genomic Map