Assay Details
Target Gene Details
Entrez Gene ID: | 81575 |
Gene Name: | apolipoprotein L domain containing 1 |
Gene Aliases: |
VERGE |
Location: |
Chr.12:12725917-12791466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOLD1 | NM_001130415.1 | NP_001123887.1 | ||
| BC042478.1 | AAH42478.1 | |||
| DC413327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map