Assay Details
Target Gene Details
Entrez Gene ID: | 7316 |
Gene Name: | ubiquitin C |
Gene Aliases: |
HMG20 |
Location: |
Chr.12:124911645-124915041 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBC | NM_021009.6 | NP_066289.3 | ||
| AB009010.1 | BAA23632.1 | |||
| AK026846.1 | ||||
| AK056939.1 | BAB71316.1 | |||
| AK098827.1 | ||||
| AK129749.1 | ||||
| AK291539.1 | ||||
| BC008955.2 | AAH08955.2 | |||
| BC039193.1 | AAH39193.1 | |||
| BC080583.1 | AAH80583.1 | |||
| BU195276.1 | ||||
| DB077241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560595 | Chr.12:124737381 - 125019207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map