Assay Details
Target Gene Details
Entrez Gene ID: | 28977 |
Gene Name: | mitochondrial ribosomal protein L42 |
Gene Aliases: |
HSPC204, L31MT, L42MT, MRP-L31, MRP-L42, MRP-S32, MRPL31, MRPS32, PTD007, RPML31, S32MT |
Location: |
Chr.12:93467490-93503772 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL42 | NM_014050.3 | 6 | 814 | NP_054769.1 |
| NM_172177.3 | 6 | 817 | NP_751917.1 | |
| NR_038159.1 | 7 | 837 | ||
| NR_038160.1 | 7 | 834 | ||
| NR_038161.1 | 7 | 919 | ||
| AF078860.1 | 6 | 796 | AAD44492.1 | |
| AF135160.1 | AAF43784.1 | |||
| AF151038.1 | 6 | 788 | AAF36124.1 | |
| AK000285.1 | 6 | 790 | BAA91054.1 | |
| AL136659.1 | 6 | 791 | CAB66594.1 | |
| BC040240.2 | 5 | 753 | AAH40240.1 | |
| CR749344.1 | 7 | 891 | CAH18197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv9445 | Chr.12:93500779 - 93502148 on Build GRCh38 | Loss |
|
| nsv1070121 | Chr.12:93501075 - 93501966 on Build GRCh38 | Deletion |
|
| esv3549592 | Chr.12:93501027 - 93502072 on Build GRCh38 | Deletion |
|
| esv3630401 | Chr.12:93501038 - 93501927 on Build GRCh38 | Loss |
|
| esv3630400 | Chr.12:93492467 - 93523879 on Build GRCh38 | Loss |
|
| esv2746217 | Chr.12:93500985 - 93501900 on Build GRCh38 | Deletion |
|
| dgv305e199 | Chr.12:93492450 - 93524021 on Build GRCh38 | Deletion |
|
| nsv1111077 | Chr.12:93500780 - 93501896 on Build GRCh38 | Tandem Duplication |
|
| nsv957426 | Chr.12:93501060 - 93501883 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map