Assay Details
Target Gene Details
Entrez Gene ID: | 5288 |
Gene Name: | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma |
Gene Aliases: |
PI3K-C2-gamma, PI3K-C2GAMMA |
Location: |
Chr.12:18242946-18716861 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3C2G | XM_011520696.2 | XP_011518998.1 | ||
| XM_017019470.1 | XP_016874959.1 | |||
| XM_017019471.1 | XP_016874960.1 | |||
| XM_017019472.1 | XP_016874961.1 | |||
| XM_017019473.1 | XP_016874962.1 | |||
| XM_017019474.1 | XP_016874963.1 | |||
| XM_017019476.1 | XP_016874965.1 | |||
| XM_017019477.1 | XP_016874966.1 | |||
| XM_017019479.1 | XP_016874968.1 |
Target Gene Details
Entrez Gene ID: | 89869 |
Gene Name: | phospholipase C zeta 1 |
Gene Aliases: |
NYD-SP27, PLC-zeta-1, PLCzeta |
Location: |
Chr.12:18645529-18738059 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCZ1 | XM_017020176.1 | 15 | 5072 | XP_016875665.1 |
| XM_017020177.1 | 16 | 4949 | XP_016875666.1 | |
| XM_017020178.1 | 14 | 4922 | XP_016875667.1 | |
| XM_017020179.1 | 15 | 5067 | XP_016875668.1 | |
| XM_017020180.1 | 15 | 4961 | XP_016875669.1 | |
| XM_017020181.1 | 15 | 5026 | XP_016875670.1 | |
| XM_017020182.1 | 11 | 4725 | XP_016875671.1 | |
| XM_017020183.1 | 12 | 4489 | XP_016875672.1 | |
| XM_017020184.1 | 12 | 4417 | XP_016875673.1 | |
| XM_017020185.1 | 11 | 4302 | XP_016875674.1 | |
| XM_017020186.1 | 13 | 4324 | XP_016875675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557672 | Chr.12:18654397 - 18698600 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74604703] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map