Assay Details
Target Gene Details
Entrez Gene ID: | 51266 |
Gene Name: | C-type lectin domain family 1 member B |
Gene Aliases: |
1810061I13Rik, CLEC2, CLEC2B, PRO1384, QDED721 |
Location: |
Chr.12:9986119-10001894 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC1B | XM_005253382.4 | XP_005253439.1 | ||
| XM_011520685.2 | XP_011518987.1 | |||
| BC029554.1 | AAH29554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1374n100 | Chr.12:9968689 - 10026775 on Build GRCh38 | Gain |
|
| nsv525593 | Chr.12:9968748 - 10004047 on Build GRCh38 | Loss |
|
| nsv604 | Chr.12:9991440 - 10007841 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map