Assay Details
Target Gene Details
Entrez Gene ID: | 26127 |
Gene Name: | FGFR1 oncogene partner 2 |
Gene Aliases: |
HSPC123-like, WIT3.0 |
Location: |
Chr.12:26938372-26966648 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR1OP2 | NM_001171887.1 | NP_001165358.1 | ||
| NM_015633.2 | NP_056448.1 | |||
| AF161472.1 | AAF29087.1 | |||
| AK001534.1 | BAA91745.1 | |||
| AK094888.1 | 1 | 200 | ||
| AL117608.1 | CAB56012.1 | |||
| AY506561.1 | AAR91611.1 | |||
| BI829470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892164 | Chr.12:26945269 - 26971722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map