Assay Details
Target Gene Details
Entrez Gene ID: | 283314 |
Gene Name: | C1RL antisense RNA 1 |
Gene Aliases: |
MATL2963 |
Location: |
Chr.12:7108308-7121851 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1RL-AS1 | NR_026947.1 | 8 | 2557 | |
| AK091792.1 | ||||
| AL833364.1 | 8 | 2557 |

Genomic Map