Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Gene Details
Entrez Gene ID: | 55703 |
Gene Name: | RNA polymerase III subunit B |
Gene Aliases: |
C128, HLD8, INMAP, RPC2 |
Location: |
Chr.12:106357658-106510198 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3B | NM_001160708.1 | NP_001154180.1 | ||
| NM_018082.5 | NP_060552.4 | |||
| XM_017019621.1 | 26 | 3291 | XP_016875110.1 | |
| AK001161.1 | BAA91527.1 | |||
| AK001250.1 | BAA91581.1 | |||
| AK090636.1 | ||||
| AK092031.1 | ||||
| AK122713.1 | ||||
| AK291635.1 | ||||
| AY092084.1 | AAM18214.1 | |||
| BC046238.1 | AAH46238.1 | |||
| BX537447.1 | CAD97689.1 | |||
| KJ818092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832510 | Chr.12:106393952 - 106557362 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map