Assay Details
Target Gene Details
Entrez Gene ID: | 8630 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 6 |
Gene Aliases: |
HSE, RODH, SDR9C6 |
Location: |
Chr.12:56752449-56787790 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B6 | NM_003725.3 | NP_003716.2 | ||
| XM_005269207.1 | XP_005269264.1 | |||
| XM_005269208.1 | XP_005269265.1 | |||
| XM_005269209.1 | XP_005269266.1 | |||
| XM_006719672.1 | XP_006719735.1 | |||
| XM_011538925.1 | XP_011537227.1 | |||
| XM_011538926.1 | XP_011537228.1 | |||
| XM_011538927.1 | XP_011537229.1 | |||
| AF223225.1 | AAF81017.1 | |||
| AK313423.1 | ||||
| BC020710.1 | AAH20710.1 | |||
| DA086202.1 | ||||
| U89281.1 | AAB88252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| esv3629600 | Chr.12:56762971 - 56780702 on Build GRCh38 | Loss |
|
| esv3580281 | Chr.12:56766287 - 56769532 on Build GRCh38 | Loss |
|
| nsv826382 | Chr.12:56757680 - 56814823 on Build GRCh38 | Gain |
|
| esv2667772 | Chr.12:56763048 - 56780839 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map