Assay Details
Target Gene Details
Entrez Gene ID: | 10443 |
Gene Name: | NEDD4 binding protein 2 like 2 |
Gene Aliases: |
92M18.3, CG005, CG016, PFAAP5 |
Location: |
Chr.13:32432488-32542710 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP2L2 | NM_001278432.1 | NP_001265361.1 | ||
| NM_001320836.1 | NP_001307765.1 | |||
| NM_033111.4 | NP_149102.3 | |||
| XM_011534869.2 | XP_011533171.1 | |||
| XM_011534870.2 | XP_011533172.1 | |||
| XM_011534871.2 | XP_011533173.1 | |||
| XM_011534872.2 | XP_011533174.1 | |||
| XM_017020328.1 | XP_016875817.1 | |||
| XM_017020329.1 | XP_016875818.1 | |||
| XM_017020330.1 | XP_016875819.1 | |||
| XM_017020333.1 | XP_016875822.1 | |||
| XM_017020334.1 | XP_016875823.1 | |||
| XM_017020335.1 | XP_016875824.1 | |||
| AK093514.1 | ||||
| AK298546.1 | ||||
| AK310280.1 | ||||
| AL049802.1 | CAB42563.1 | |||
| BC131631.1 | ||||
| CB240991.1 | ||||
| U50529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832576 | Chr.13:32322994 - 32502851 on Build GRCh38 | Gain |
|
| esv2747243 | Chr.13:32433429 - 32436635 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map