Assay Details
Target Gene Details
Entrez Gene ID: | 8600 |
Gene Name: | tumor necrosis factor superfamily member 11 |
Gene Aliases: |
CD254, ODF, OPGL, OPTB2, RANKL, TNLG6B, TRANCE, hRANKL2, sOdf |
Location: |
Chr.13:42562736-42608013 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF11 | NM_033012.3 | NP_143026.1 |
More Information
Set Membership: |
|

Genomic Map