Assay Details
Target Gene Details
Entrez Gene ID: | 122042 |
Gene Name: | relaxin/insulin like family peptide receptor 2 |
Gene Aliases: |
GPR106, GREAT, INSL3R, LGR8, LGR8.1, RXFPR2 |
Location: |
Chr.13:31739542-31802872 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RXFP2 | NM_001166058.1 | NP_001159530.1 | ||
| NM_130806.3 | NP_570718.1 | |||
| XM_017020389.1 | XP_016875878.1 | |||
| AF403384.2 | AAL69324.2 | |||
| AF453828.1 | AAL73946.1 | |||
| AY899851.1 | AAX85199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580592 | Chr.13:31762523 - 31770852 on Build GRCh38 | Loss |
|
| dgv1625n100 | Chr.13:31678835 - 31773086 on Build GRCh38 | Loss |
|
| nsv561401 | Chr.13:31682595 - 31771071 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map