Assay Details
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
| NM_001202559.1 | NP_001189488.1 | |||
| AK296850.1 | ||||
| AK303739.1 |
Target Gene Details
Entrez Gene ID: | 376267 |
Gene Name: | RAB15, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.14:64945814-64972197 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB15 | NM_001308154.1 | NP_001295083.1 | ||
| NM_198686.2 | NP_941959.1 | |||
| XM_017021277.1 | XP_016876766.1 | |||
| AB672631.2 | ||||
| AB672632.1 | ||||
| AB678452.1 | ||||
| AB678453.1 | ||||
| BC040679.2 | AAH40679.2 | |||
| BX248046.1 | CAD62353.1 | |||
| BX640825.1 | CAE45901.2 | |||
| CN352234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map