Assay Details
Target Gene Details
Entrez Gene ID: | 646658 |
Gene Name: | synapse differentiation inducing 1 like |
Gene Aliases: |
CAPUCIN, DSPC1, IFITMD4, TMEM90A |
Location: |
Chr.14:74405893-74477080 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNDIG1L | XM_017021600.1 | XP_016877089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634943 | Chr.14:74426684 - 74537774 on Build GRCh38 | Gain |
|
| nsv832829 | Chr.14:74377797 - 74545100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map