Assay Details
Target Gene Details
Entrez Gene ID: | 4329 |
Gene Name: | aldehyde dehydrogenase 6 family member A1 |
Gene Aliases: |
MMSADHA, MMSDH |
Location: |
Chr.14:74057662-74084493 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH6A1 | NM_001278593.1 | 12 | 3114 | NP_001265522.1 |
| NM_001278594.1 | 12 | 3316 | NP_001265523.1 | |
| NM_005589.3 | 12 | 3153 | NP_005580.1 | |
| XM_017021331.1 | 10 | 3004 | XP_016876820.1 |
Target Gene Details
Entrez Gene ID: | 80127 |
Gene Name: | basal body orientation factor 1 |
Gene Aliases: |
C14orf45, CCDC176 |
Location: |
Chr.14:74019350-74078351 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOF1 | NM_025057.2 | NP_079333.2 | ||
| XM_011537171.2 | XP_011535473.1 | |||
| XM_011537172.1 | XP_011535474.1 | |||
| XM_011537174.2 | XP_011535476.1 | |||
| XM_017021659.1 | XP_016877148.1 | |||
| XM_017021660.1 | XP_016877149.1 | |||
| XM_017021661.1 | XP_016877150.1 | |||
| XM_017021662.1 | XP_016877151.1 | |||
| XM_017021663.1 | XP_016877152.1 | |||
| AK026842.1 | BAB15572.1 | |||
| AK097531.1 | ||||
| AL834487.1 | CAD39145.1 | |||
| BC104979.1 | AAI04980.1 | |||
| BC104981.1 | AAI04982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634932 | Chr.14:74053535 - 74063579 on Build GRCh38 | Loss |
|
| dgv1936n100 | Chr.14:73950133 - 74073431 on Build GRCh38 | Gain |
|
| dgv1938n100 | Chr.14:73984364 - 74075786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map