Assay Details
Target Gene Details
Entrez Gene ID: | 51222 |
Gene Name: | zinc finger protein 219 |
Gene Aliases: |
ZFP219 |
Location: |
Chr.14:21090046-21104704 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF219 | NM_001101672.1 | NP_001095142.1 | ||
| NM_001102454.1 | NP_001095924.1 | |||
| NM_016423.2 | NP_057507.2 | |||
| XM_005267739.1 | XP_005267796.1 | |||
| XM_006720163.2 | XP_006720226.1 | |||
| XM_006720164.3 | XP_006720227.1 | |||
| XM_017021355.1 | XP_016876844.1 | |||
| AB015427.1 | BAA90526.1 | |||
| AK225811.1 | ||||
| AK290700.1 | ||||
| BC036105.1 | AAH36105.1 | |||
| CB152705.1 | ||||
| DA013397.1 | ||||
| DC358466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv123n21 | Chr.14:21072704 - 21106531 on Build GRCh38 | Loss |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map