Assay Details
Target Gene Details
Entrez Gene ID: | 145474 |
Gene Name: | uncharacterized LOC145474 |
Gene Aliases: |
- |
Location: |
Chr.14:71487861-71489714 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC145474 | NR_027046.1 | 1 | 1052 | |
| AK023718.1 | 1 | 1052 | ||
| BX102012.1 | 1 | 261 |
Target Gene Details
Entrez Gene ID: | 26037 |
Gene Name: | signal induced proliferation associated 1 like 1 |
Gene Aliases: |
E6TP1 |
Location: |
Chr.14:71320418-71741229 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1341 | Chr.14:71481869 - 71513924 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map