Assay Details
Target Gene Details
Entrez Gene ID: | 729665 |
Gene Name: | coiled-coil domain containing 175 |
Gene Aliases: |
C14orf38, c14_5395 |
Location: |
Chr.14:59504212-59577276 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC175 | NM_001164399.1 | NP_001157871.1 | ||
| XM_011537120.2 | 20 | 3059 | XP_011535422.1 | |
| XM_011537122.2 | 18 | 2836 | XP_011535424.1 | |
| XM_011537124.2 | 17 | 2683 | XP_011535426.1 |
Target Gene Details
Entrez Gene ID: | 51528 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
C14orf100, CDA06, HSPC213, HSPC327, JAMP |
Location: |
Chr.14:59484443-59505407 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JKAMP | NM_001098625.2 | 7 | 2051 | NP_001092095.1 |
| NM_001284201.1 | 7 | 2592 | NP_001271130.1 | |
| NM_001284202.1 | 8 | 2178 | NP_001271131.1 | |
| NM_001284203.1 | 7 | 2101 | NP_001271132.1 | |
| NM_001284204.1 | 6 | 1992 | NP_001271133.1 | |
| NM_016475.4 | 7 | 2069 | NP_057559.2 | |
| AA461198.1 | 1 | 309 | ||
| HY340214.1 | 1 | 379 |
More Information
Set Membership: |
|

Genomic Map