Assay Details
Target Gene Details
Entrez Gene ID: | 100506237 |
Gene Name: | NKX2-1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:36519278-36523016 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NKX2-1-AS1 | NR_103710.1 | |||
BX161496.1 | CAD61942.1 | |||
DA256249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv23132 | Chr.14:36517126 - 36521753 on Build GRCh38 | Loss |
![]() ![]() |
