Assay Details
Target Gene Details
Entrez Gene ID: | 8110 |
Gene Name: | double PHD fingers 3 |
Gene Aliases: |
BAF45C, CERD4 |
Location: |
Chr.14:72609032-72894116 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPF3 | XM_017021670.1 | 11 | 2074 | XP_016877159.1 |
| AK024141.1 | BAB14838.1 |
Target Gene Details
Entrez Gene ID: | 9628 |
Gene Name: | regulator of G-protein signaling 6 |
Gene Aliases: |
GAP |
Location: |
Chr.14:71932100-72632478 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS6 | XM_017021825.1 | XP_016877314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565049 | Chr.14:72603947 - 72656563 on Build GRCh38 | Loss |
|
| nsv456337 | Chr.14:72595825 - 72622005 on Build GRCh38 | Loss |
|
| esv3892558 | Chr.14:72255497 - 72756402 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map