Assay Details
Target Gene Details
Entrez Gene ID: | 122830 |
Gene Name: | N(alpha)-acetyltransferase 30, NatC catalytic subunit |
Gene Aliases: |
C14orf35, MAK3, Mak3p, NAT12, NAT12P |
Location: |
Chr.14:57390553-57412748 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA30 | NM_001011713.2 | NP_001011713.2 | ||
| AK092674.1 | ||||
| AK296372.1 | ||||
| AK304257.1 | ||||
| AK307348.1 | ||||
| BC118589.1 | ||||
| BC122557.1 | ||||
| BX648700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046422 | Chr.14:57198718 - 58129812 on Build GRCh38 | Gain |
|
| nsv832806 | Chr.14:57284961 - 57488279 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113320757] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map