Assay Details
Target Gene Details
Entrez Gene ID: | 5826 |
Gene Name: | ATP binding cassette subfamily D member 4 |
Gene Aliases: |
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L |
Location: |
Chr.14:74285277-74303064 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565183 | Chr.14:74265506 - 74292774 on Build GRCh38 | Loss |
|
| nsv565182 | Chr.14:74220714 - 74299190 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map