Assay Details
Target Gene Details
Entrez Gene ID: | 122769 |
Gene Name: | leucine rich repeat protein 1 |
Gene Aliases: |
4-1BBLRR, LRR-1, PPIL5 |
Location: |
Chr.14:49598697-49614672 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRR1 | NM_152329.3 | NP_689542.2 | ||
| NM_203467.1 | NP_982292.1 | |||
| NR_037792.1 | ||||
| NR_037793.1 | ||||
| AA742244.1 | ||||
| AK293156.1 | ||||
| AY052405.1 | AAL11430.1 | |||
| BC030142.2 | AAH30142.1 | |||
| BC064133.1 | AAH64133.1 | |||
| BC093697.1 | AAH93697.1 | |||
| BC107888.1 | AAI07889.1 | |||
| BC112241.1 | AAI12242.1 | |||
| BC139921.1 | ||||
| BQ059807.1 | ||||
| BX248298.1 | CAD62625.1 | |||
| BX648029.1 | CAH10374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map