Assay Details
Target Gene Details
Entrez Gene ID: | 145389 |
Gene Name: | solute carrier family 38 member 6 |
Gene Aliases: |
NAT-1, SNAT6 |
Location: |
Chr.14:60981114-61083733 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A6 | NM_001172702.1 | NP_001166173.1 | ||
| NM_153811.2 | NP_722518.2 | |||
| NR_033344.1 | ||||
| XM_006720049.3 | XP_006720112.1 | |||
| XM_011536469.2 | XP_011534771.1 | |||
| XM_017021020.1 | XP_016876509.1 | |||
| XM_017021021.1 | XP_016876510.1 | |||
| XM_017021022.1 | XP_016876511.1 | |||
| XM_017021023.1 | XP_016876512.1 | |||
| XM_017021024.1 | XP_016876513.1 | |||
| XM_017021025.1 | XP_016876514.1 | |||
| XM_017021026.1 | XP_016876515.1 | |||
| AF070578.1 | ||||
| AF543422.1 | AAN47144.1 | |||
| AK303839.1 | ||||
| BC110378.1 | ||||
| BP352279.1 | ||||
| BX248072.1 | CAD62361.1 | |||
| BX648332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037387 | Chr.14:60742756 - 61066784 on Build GRCh38 | Gain |
|
| esv2748755 | Chr.14:60887693 - 61016326 on Build GRCh38 | Deletion |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
| nsv1049936 | Chr.14:60934305 - 61080658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map