Assay Details
Target Gene Details
Entrez Gene ID: | 646548 |
Gene Name: | ADAM metallopeptidase domain 20 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.14:70187108-70232583 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC646548 | NR_135825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832826 | Chr.14:70092872 - 70256747 on Build GRCh38 | Gain |
|

Genomic Map