Assay Details
Target Gene Details
Entrez Gene ID: | 51527 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
C14orf129, HSPC210 |
Location: |
Chr.14:96363452-96387290 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSKIP | NM_001271904.1 | NP_001258833.1 | ||
| NM_001271905.1 | NP_001258834.1 | |||
| NM_001271906.1 | NP_001258835.1 | |||
| NM_016472.4 | NP_057556.2 | |||
| XM_017021365.1 | XP_016876854.1 | |||
| AF151044.1 | AAF36130.1 | |||
| AK000796.1 | BAA91380.1 | |||
| AK094654.1 | ||||
| AK314423.1 | ||||
| BC004818.2 | AAH04818.1 | |||
| DA488110.1 | ||||
| HY019704.1 | ||||
| HY051680.1 | ||||
| HY108584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565649 | Chr.14:96377253 - 96453283 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114610244] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map