Assay Details
Target Gene Details
Entrez Gene ID: | 2954 |
Gene Name: | glutathione S-transferase zeta 1 |
Gene Aliases: |
GSTZ1-1, MAAI, MAI |
Location: |
Chr.14:77320887-77331597 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTZ1 | NM_001312660.1 | NP_001299589.1 | ||
| NM_145870.2 | NP_665877.1 | |||
| NM_145871.2 | NP_665878.2 | |||
| XM_011536670.2 | XP_011534972.1 | |||
| XM_011536671.2 | XP_011534973.1 | |||
| XM_017021243.1 | XP_016876732.1 | |||
| AB209360.1 | BAD92597.1 | |||
| AJ001838.1 | CAA05045.1 | |||
| AK315154.1 | ||||
| AK315609.1 | ||||
| BC001453.1 | AAH01453.1 | |||
| BG330365.1 | ||||
| BG772450.1 | ||||
| BI559916.1 | ||||
| CR456987.1 | CAG33268.1 | |||
| U86529.1 | AAB96392.1 |
More Information
Set Membership: |
|

Genomic Map