Assay Details
Target Gene Details
Entrez Gene ID: | 101928909 |
Gene Name: | uncharacterized LOC101928909 |
Gene Aliases: |
- |
Location: |
Chr.14:90642629-90648891 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928909 | NR_110134.1 | 2 | 187 | |
| HY025890.1 | 2 | 188 |
Target Gene Details
Entrez Gene ID: | 145567 |
Gene Name: | tetratricopeptide repeat domain 7B |
Gene Aliases: |
TTC7L1, c14_5685 |
Location: |
Chr.14:90540588-90816479 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC7B | NM_001010854.1 | NP_001010854.1 | ||
| NM_001320421.1 | NP_001307350.1 | |||
| XM_005267367.1 | XP_005267424.1 | |||
| XM_011536497.1 | XP_011534799.1 | |||
| XM_017021044.1 | XP_016876533.1 | |||
| XM_017021045.1 | XP_016876534.1 | |||
| XM_017021046.1 | XP_016876535.1 | |||
| XM_017021047.1 | XP_016876536.1 | |||
| AK126571.1 | ||||
| AL832848.1 | CAI46128.1 | |||
| BC035865.2 | AAH35865.1 | |||
| BC048270.2 | AAH48270.2 | |||
| BX248275.1 | CAD62603.1 |
More Information
Set Membership: |
|

Genomic Map