Assay Details
Target Gene Details
Entrez Gene ID: | 123041 |
Gene Name: | solute carrier family 24 member 4 |
Gene Aliases: |
AI2A5, NCKX4, SHEP6, SLC24A2 |
Location: |
Chr.14:92322581-92501481 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A4 | NM_153646.3 | NP_705932.2 | ||
| NM_153647.3 | NP_705933.2 | |||
| NM_153648.3 | NP_705934.1 | |||
| XM_011536436.2 | XP_011534738.1 | |||
| XM_011536437.2 | XP_011534739.1 | |||
| XM_011536438.2 | XP_011534740.1 | |||
| XM_011536439.2 | XP_011534741.1 | |||
| AF520704.1 | AAM76070.1 | |||
| AF520705.1 | AAM76071.1 | |||
| AF520706.1 | AAM76072.1 | |||
| AK295059.1 | ||||
| BC069653.1 | AAH69653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635338 | Chr.14:92333836 - 92336196 on Build GRCh38 | Loss |
|
| esv2678086 | Chr.14:92333878 - 92336226 on Build GRCh38 | Deletion |
|
| nsv832859 | Chr.14:92281645 - 92449899 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map