Assay Details
Target Gene Details
Entrez Gene ID: | 646548 |
Gene Name: | ADAM metallopeptidase domain 20 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.14:70187108-70232583 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC646548 | NR_135825.1 |
Target Gene Details
Entrez Gene ID: | 6547 |
Gene Name: | solute carrier family 8 member A3 |
Gene Aliases: |
NCX3 |
Location: |
Chr.14:70044217-70189626 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8A3 | NM_033262.4 | NP_150287.1 | ||
| NM_058240.3 | NP_489479.1 | |||
| NM_182932.2 | NP_891977.1 | |||
| NM_183002.2 | NP_892114.1 | |||
| NR_104122.1 | ||||
| XM_017021606.1 | XP_016877095.1 | |||
| XM_017021607.1 | XP_016877096.1 | |||
| XM_017021608.1 | XP_016877097.1 | |||
| XM_017021609.1 | XP_016877098.1 | |||
| XM_017021610.1 | XP_016877099.1 | |||
| AF510501.1 | AAN60790.1 | |||
| AF510502.1 | AAN60791.1 | |||
| AF510503.1 | AAN60792.1 | |||
| BI906774.1 | ||||
| BX248763.1 | CAD66570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1338 | Chr.14:70165006 - 70197290 on Build GRCh38 | Insertion |
|
| nsv832826 | Chr.14:70092872 - 70256747 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map