Assay Details
Target Gene Details
Entrez Gene ID: | 64841 |
Gene Name: | glucosamine-phosphate N-acetyltransferase 1 |
Gene Aliases: |
GNA1, GNPNAT, Gpnat1 |
Location: |
Chr.14:52775193-52791668 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNPNAT1 | NM_198066.3 | NP_932332.1 | ||
| XM_005268012.2 | XP_005268069.1 | |||
| XM_006720238.3 | XP_006720301.1 | |||
| AK090577.1 | BAC03482.1 | |||
| BC012179.1 | AAH12179.1 | |||
| BG717628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv7884 | Chr.14:52785633 - 52788584 on Build GRCh38 | Loss |
|
| nsv832796 | Chr.14:52680158 - 52860182 on Build GRCh38 | Loss |
|
| nsv819339 | Chr.14:52785718 - 52788066 on Build GRCh38 | Loss |
|
| nsv1050582 | Chr.14:52773715 - 52861657 on Build GRCh38 | Gain |
|
| nsv826956 | Chr.14:52785859 - 52788244 on Build GRCh38 | Loss |
|
| nsv1279 | Chr.14:52754853 - 52788471 on Build GRCh38 | Insertion |
|
| esv3634528 | Chr.14:52785748 - 52788377 on Build GRCh38 | Loss |
|
| nsv470647 | Chr.14:52730993 - 52831559 on Build GRCh38 | Gain |
|
| esv2748720 | Chr.14:52785733 - 52788459 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map