Assay Details
Target Gene Details
Entrez Gene ID: | 8747 |
Gene Name: | ADAM metallopeptidase domain 21 |
Gene Aliases: |
ADAM 21, ADAM31 |
Location: |
Chr.14:70452157-70459905 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM21 | NM_003813.3 | NP_003804.2 | ||
| DB035527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581280 | Chr.14:70442517 - 70472933 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map