Assay Details
Target Gene Details
Entrez Gene ID: | 400242 |
Gene Name: | DICER1 antisense RNA 1 |
Gene Aliases: |
DICER1-AS |
Location: |
Chr.14:95157688-95179933 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DICER1-AS1 | NR_015415.1 | |||
| BI754942.1 | ||||
| BX648898.1 | 1 | 3514 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036738 | Chr.14:95140842 - 95292222 on Build GRCh38 | Gain |
|
| esv3635396 | Chr.14:95103520 - 95288202 on Build GRCh38 | Gain |
|

Genomic Map