Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 10490 |
Gene Name: | vesicle transport through interaction with t-SNAREs 1B |
Gene Aliases: |
VTI1, VTI1-LIKE, VTI1L, VTI2, v-SNARE, vti1-rp1 |
Location: |
Chr.14:67651149-67674885 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VTI1B | NM_006370.2 | NP_006361.1 | ||
| AF035824.1 | AAC52016.1 | |||
| AF060902.1 | AAC73059.1 | |||
| AK291689.1 | ||||
| AK310839.1 | ||||
| AK316572.1 | ||||
| BC003142.1 | AAH03142.1 | |||
| BT019348.1 | AAV38155.1 | |||
| BT019960.1 | AAV38763.1 | |||
| CR456757.1 | CAG33038.1 | |||
| CR542095.1 | CAG46892.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73274582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map