Assay Details
Target Gene Details
Entrez Gene ID: | 80224 |
Gene Name: | nucleotide binding protein like |
Gene Aliases: |
C14orf127, IND1, huInd1 |
Location: |
Chr.14:31561385-31861293 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUBPL | NM_001201573.1 | NP_001188502.1 | ||
| NM_025152.2 | NP_079428.2 | |||
| NR_120408.1 | ||||
| XM_011537181.2 | XP_011535483.1 | |||
| XM_011537182.2 | XP_011535484.1 | |||
| XM_011537183.2 | XP_011535485.1 | |||
| XM_017021664.1 | XP_016877153.1 | |||
| XM_017021665.1 | XP_016877154.1 | |||
| XM_017021666.1 | XP_016877155.1 | |||
| XM_017021667.1 | XP_016877156.1 | |||
| AK022722.1 | BAB14203.1 | |||
| AK295326.1 | ||||
| AK301447.1 | ||||
| AK316445.1 | ||||
| BC024919.1 | AAH24919.1 | |||
| BP274976.1 | ||||
| BX248028.1 | CAD62349.1 | |||
| KJ534907.1 | ||||
| KJ534908.1 | ||||
| KJ534909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3607n54 | Chr.14:31489360 - 31899728 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map