Assay Details
Target Gene Details
Entrez Gene ID: | 2287 |
Gene Name: | FK506 binding protein 3 |
Gene Aliases: |
FKBP-25, FKBP-3, FKBP25, PPIase |
Location: |
Chr.14:45115599-45134529 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP3 | NM_002013.3 | NP_002004.1 | ||
| XM_011536565.2 | XP_011534867.1 | |||
| XM_017021092.1 | XP_016876581.1 | |||
| XM_017021093.1 | XP_016876582.1 | |||
| AK222993.1 | BAD96713.1 | |||
| AK311915.1 | ||||
| BC016288.1 | AAH16288.1 | |||
| BC020809.1 | AAH20809.1 | |||
| BI598048.1 | ||||
| BT006904.1 | AAP35550.1 | |||
| M96256.1 | AAA58471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map