Assay Details
Target Gene Details
Entrez Gene ID: | 100506237 |
Gene Name: | NKX2-1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:36519278-36523016 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-1-AS1 | NR_103710.1 | 2 | 682 | |
| BX161496.1 | 2 | 644 | CAD61942.1 |

Genomic Map