Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLC1 | NM_001130107.1 | NP_001123579.1 | ||
| NM_005552.4 | NP_005543.2 | |||
| NM_182923.3 | NP_891553.2 | |||
| AF527736.1 | AAP85635.1 | |||
| AK294508.1 | ||||
| AY180164.2 | AAO62549.1 | |||
| BC008881.2 | AAH08881.1 | |||
| BP274336.1 | ||||
| DA501532.1 | ||||
| L04733.1 | AAA16576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832883 | Chr.14:103461350 - 103654616 on Build GRCh38 | Loss |
|
| esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain |
|
| nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map