Assay Details
Target Gene Details
Entrez Gene ID: | 80127 |
Gene Name: | basal body orientation factor 1 |
Gene Aliases: |
C14orf45, CCDC176 |
Location: |
Chr.14:74019350-74078351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOF1 | NM_025057.2 | NP_079333.2 | ||
| XM_005268092.3 | XP_005268149.1 | |||
| XM_005268097.3 | XP_005268154.1 | |||
| XM_005268098.4 | XP_005268155.1 | |||
| XM_011537170.2 | XP_011535472.1 | |||
| XM_011537171.2 | XP_011535473.1 | |||
| XM_011537172.1 | XP_011535474.1 | |||
| XM_011537174.2 | XP_011535476.1 | |||
| XM_011537175.2 | XP_011535477.1 | |||
| XM_011537176.2 | XP_011535478.1 | |||
| XM_011537177.2 | XP_011535479.1 | |||
| XM_011537179.2 | XP_011535481.1 | |||
| XM_017021659.1 | XP_016877148.1 | |||
| XM_017021660.1 | XP_016877149.1 | |||
| XM_017021661.1 | XP_016877150.1 | |||
| XM_017021662.1 | XP_016877151.1 | |||
| XM_017021663.1 | XP_016877152.1 | |||
| DB484577.1 | ||||
| DB493409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv1936n100 | Chr.14:73950133 - 74073431 on Build GRCh38 | Gain |
|
| dgv1938n100 | Chr.14:73984364 - 74075786 on Build GRCh38 | Gain |
|
| nsv1046028 | Chr.14:73957079 - 74032042 on Build GRCh38 | Loss |
|
| nsv524396 | Chr.14:73958235 - 74023032 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map