Assay Details
Target Gene Details
Entrez Gene ID: | 10538 |
Gene Name: | basic leucine zipper ATF-like transcription factor |
Gene Aliases: |
B-ATF, BATF1, SFA-2, SFA2 |
Location: |
Chr.14:75522441-75546992 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BATF | NM_006399.3 | NP_006390.1 | ||
| BC032294.1 | AAH32294.1 | |||
| D42106.1 | BAA07686.1 | |||
| U15460.1 | AAC50314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037822 | Chr.14:75468606 - 75633843 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78749365] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map