Assay Details
Target Gene Details
Entrez Gene ID: | 102724360 |
Gene Name: | uncharacterized LOC102724360 |
Gene Aliases: |
- |
Location: |
Chr.19:18940254-18946831 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724360 | NR_134912.1 | 3 | 571 | |
| NR_134913.1 | 3 | 567 | ||
| BF511074.1 | 1 | 475 | ||
| CF127232.1 | 3 | 241 | ||
| DA366162.1 | 3 | 500 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|

Genomic Map