Assay Details
Target Gene Details
Entrez Gene ID: | 126328 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A11 |
Gene Aliases: |
B14.7, CI-B14.7 |
Location: |
Chr.19:5891276-5904013 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA11 | NM_001193375.1 | NP_001180304.1 | ||
| AK127692.1 | BAC87088.1 | |||
| BQ955016.1 |
More Information
Set Membership: |
|

Genomic Map